Recombinat Protein,Human LILRB2 CD85d ILT4 Protein, His Tag,Human LILRB2, His Tag (LI2-H5220) is expressed from human 293 cells (HEK 293). It contains AA Gln 22 - Val 461 (Accession AAH36827).This protein carries a polyhistidine tag at the C-terminus 1 * 100 µG
Catalog # ACRBLI2-H5220100UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together