Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Mouse GPA33 A33 Protein, His Tag,Mouse GPA33, His Tag (GP3-M5225) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Ile 235 (Accession NP067623.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBGP3-M5225-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse GPA33 A33 Protein, His Tag,Mouse GPA33, His Tag (GP3-M5225) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Ile 235 (Accession NP067623.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBGP3-M5225-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together