Order Entry
Ireland
ContactUsLinkComponent
Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession Q9QZF2- 1). 1 * 100 µG
Catalog # ACRBGP1-M52H7100UG
Supplier:  ACRO Biosystems
Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession Q9QZF2- 1). 1 * 100 µG
Catalog # ACRBGP1-M52H7100UG
Supplier:  ACRO Biosystems
Frequently Bought Together