Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (GP3-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP005594665.1). 1 * 100 µG
Catalog # ACRBGP3-C5259100UG
Supplier:  ACRO Biosystems
Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (GP3-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession XP005594665.1). 1 * 100 µG
Catalog # ACRBGP3-C5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together