Order Entry
Ireland
ContactUsLinkComponent
Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1) . 1 * 1 mg
Catalog # ACRBSI9-H52H4-1MG
Supplier:  ACROBIOSYSTEMS
Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1) . 1 * 1 mg
Catalog # ACRBSI9-H52H4-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together