Order Entry
Ireland
ContactUsLinkComponent
Human GITR, His Tag (GIR-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession Q9Y5U5-1). 1 * 1 mg
Catalog # ACRBGIR-H5228-1MG
Supplier:  ACRO Biosystems
Human GITR, His Tag (GIR-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession Q9Y5U5-1). 1 * 1 mg
Catalog # ACRBGIR-H5228-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together