Order Entry
Ireland
ContactUsLinkComponent
Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835) . 1 * 100 µG
Catalog # ACRBTR1-H5228100UG
Supplier:  ACRO Biosystems
Human TRAIL R1, His Tag (TR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession NP003835) . 1 * 100 µG
Catalog # ACRBTR1-H5228100UG
Supplier:  ACRO Biosystems
Frequently Bought Together