Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human NKp46 NCR1 CD335 Protein, His Tag,Human NKp46, His Tag (NC1-H52H4) is expressed from human 293 cells (HEK29 3). It contains AA Gln 22 - Asn 254 (Accession AAH64806.1).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Catalog # ACRBNC1-H52H4-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human NKp46 NCR1 CD335 Protein, His Tag,Human NKp46, His Tag (NC1-H52H4) is expressed from human 293 cells (HEK29 3). It contains AA Gln 22 - Asn 254 (Accession AAH64806.1).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Catalog # ACRBNC1-H52H4-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together