Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human IL-2, Fc,Avitag (IL2-H82F3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accessio n P60568-1). 1 * 25 µG
Catalog # ACRBIL2-H82F3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-2, Fc,Avitag (IL2-H82F3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accessio n P60568-1). 1 * 25 µG
Catalog # ACRBIL2-H82F3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together