Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Mouse IL-23 R Protein, Fc Tag,Mouse IL-23R, Fc Tag (ILR-M5251) is expressed from human 293 cells (HEK293). It con tains AA Gly 24 - Asp 372 (Accession NP653131).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBILR-M5251-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Mouse IL-23 R Protein, Fc Tag,Mouse IL-23R, Fc Tag (ILR-M5251) is expressed from human 293 cells (HEK293). It con tains AA Gly 24 - Asp 372 (Accession NP653131).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBILR-M5251-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together