Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human APRIL, Fc,Avitag (APL-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ala 105 - Leu 250 (Access ion O75888-1). 1 * 25 µG
Catalog # ACRBAPL-H82F5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human APRIL, Fc,Avitag (APL-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ala 105 - Leu 250 (Access ion O75888-1). 1 * 25 µG
Catalog # ACRBAPL-H82F5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together