Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human SCARB2 SR-B2 LIMPII Protein, Fc Tag,Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (HEK 293). It contains AA Arg 27 - Thr 432 (Accession AAH21892).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBSC2-H5250100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human SCARB2 SR-B2 LIMPII Protein, Fc Tag,Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (HEK 293). It contains AA Arg 27 - Thr 432 (Accession AAH21892).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBSC2-H5250100UG
Supplier:  ACRO Biosystems
Frequently Bought Together