Order Entry
Ireland
ContactUsLinkComponent
Rhesus macaque Cynomolgus Complement Factor D, His Tag (CFD-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Ala 253 (Accession H9EXC1-1). In the region Ile 26 - Ala 253, the AA sequence of Rhesus macaque and Cynomolgus Com 1 * 100 µG
Catalog # ACRBCFD-R52H3100UG
Supplier:  ACRO Biosystems
Rhesus macaque Cynomolgus Complement Factor D, His Tag (CFD-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Ala 253 (Accession H9EXC1-1). In the region Ile 26 - Ala 253, the AA sequence of Rhesus macaque and Cynomolgus Com 1 * 100 µG
Catalog # ACRBCFD-R52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together