Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human IL-27 Ra TCCR Protein, Fc Tag,Human IL-27 Ra, Fc Tag (ILA-H5254) is expressed from human 293 cells (HEK293) . It contains AA Gln 33 - Arg 514 (Accession AAH28003).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 1 mg
Catalog # ACRBILA-H5254-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human IL-27 Ra TCCR Protein, Fc Tag,Human IL-27 Ra, Fc Tag (ILA-H5254) is expressed from human 293 cells (HEK293) . It contains AA Gln 33 - Arg 514 (Accession AAH28003).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 1 mg
Catalog # ACRBILA-H5254-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together