Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human IL-5 R alpha CD125 Protein, His Tag,Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 cel ls (HEK293). It contains AA Asp 21 - Glu 335 (Accession NP000555).This protein carries a polyhistidine tag at the C-t 1 * 1 mg
Catalog # ACRBILA-H52H4-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human IL-5 R alpha CD125 Protein, His Tag,Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 cel ls (HEK293). It contains AA Asp 21 - Glu 335 (Accession NP000555).This protein carries a polyhistidine tag at the C-t 1 * 1 mg
Catalog # ACRBILA-H52H4-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together