Order Entry
Ireland
ContactUsLinkComponent
FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254). 1 * 200 µG
Catalog # ACRBBCA-HF254200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254). 1 * 200 µG
Catalog # ACRBBCA-HF254200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together