Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human SCARB1 SR-B1 Protein, Fc Tag,Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). I t contains AA Pro 33 - Tyr 443 (Accession NP005496.4).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBSC1-H5258-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human SCARB1 SR-B1 Protein, Fc Tag,Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). I t contains AA Pro 33 - Tyr 443 (Accession NP005496.4).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBSC1-H5258-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together