Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Rabbit CD47 Protein, Fc Tag,Rabbit CD47, Fc Tag (CD7-R5257) is expressed from human 293 cells (HEK293). It contai ns AA Gln 19 - Glu 139 (Accession G1SKT7).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBCD7-R5257-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Rabbit CD47 Protein, Fc Tag,Rabbit CD47, Fc Tag (CD7-R5257) is expressed from human 293 cells (HEK293). It contai ns AA Gln 19 - Glu 139 (Accession G1SKT7).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBCD7-R5257-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together