Order Entry
Ireland
ContactUsLinkComponent
Human B7-H4, Fc Tag, low endotoxin (B74-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accessio n Q7Z7D3-1). 1 * 100 µG
  ACRBB74-H5256100UG
Human B7-H4, Fc Tag, low endotoxin (B74-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accessio n Q7Z7D3-1). 1 * 100 µG
  ACRBB74-H5256100UG
Frequently Bought Together