Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human Serpin F1 PEDF Protein, His Tag,Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HEK 293). It contains AA Gln 20 - Pro 418 (Accession NP002606).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Catalog # ACRBSE1-H5221-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Serpin F1 PEDF Protein, His Tag,Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HEK 293). It contains AA Gln 20 - Pro 418 (Accession NP002606).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Catalog # ACRBSE1-H5221-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together