Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Cynomolgus LILRB4 CD85k ILT3 Protein, Mouse IgG2a Fc Tag,Cynomolgus LILRB4, Mouse IgG2a Fc Tag (LI4-C5250) is exp ressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession XP015297198.1).This protein carries 1 * 100 µG
Catalog # ACRBLI4-C5250100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Cynomolgus LILRB4 CD85k ILT3 Protein, Mouse IgG2a Fc Tag,Cynomolgus LILRB4, Mouse IgG2a Fc Tag (LI4-C5250) is exp ressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession XP015297198.1).This protein carries 1 * 100 µG
Catalog # ACRBLI4-C5250100UG
Supplier:  ACRO Biosystems
Frequently Bought Together