Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human LAG-3, Mouse IgG2a Fc,Avitag (LA3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession P18627-1). 1 * 200 µG
  ACRBLA3-H82F3200UG
Biotinylated Human LAG-3, Mouse IgG2a Fc,Avitag (LA3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession P18627-1). 1 * 200 µG
  ACRBLA3-H82F3200UG
Frequently Bought Together