Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human CD3 epsilon, His,Avitag (CDE-H82E1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 ( Accession P07766-1). 1 * 25 µG
Catalog # ACRBCDE-H82E1-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CD3 epsilon, His,Avitag (CDE-H82E1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 ( Accession P07766-1). 1 * 25 µG
Catalog # ACRBCDE-H82E1-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together