Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human M-CSF, His,Avitag (MCF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Arg 255 (Access ion P09603-1). 1 * 200 µG
Catalog # ACRBMCF-H82E6200UG
Supplier:  ACRO Biosystems
Biotinylated Human M-CSF, His,Avitag (MCF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Arg 255 (Access ion P09603-1). 1 * 200 µG
Catalog # ACRBMCF-H82E6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together