Order Entry
Ireland
ContactUsLinkComponent
Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession P15692-9). 1 * 50 µG
Catalog # ACRBVE1-H5246-50UG
Supplier:  ACRO Biosystems
Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession P15692-9). 1 * 50 µG
Catalog # ACRBVE1-H5246-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together