Order Entry
Ireland
ContactUsLinkComponent
Mouse 4-1BB, Fc Tag (41B-M5258) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Leu 211 (Accession Q8R037-1). 1 * 1 mg
Catalog # ACRB41B-M5258-1MG
Supplier:  ACRO Biosystems
Mouse 4-1BB, Fc Tag (41B-M5258) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Leu 211 (Accession Q8R037-1). 1 * 1 mg
Catalog # ACRB41B-M5258-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together