Order Entry
Ireland
ContactUsLinkComponent
FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (Ac cession P51654-1). It is the FITC labeled form of Human Glypican 3, Fc Tag (Cat. No. GP3-H5258). 1 * 200 µG
Catalog # ACRBGP3-HF258200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (Ac cession P51654-1). It is the FITC labeled form of Human Glypican 3, Fc Tag (Cat. No. GP3-H5258). 1 * 200 µG
Catalog # ACRBGP3-HF258200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together