Order Entry
Ireland
ContactUsLinkComponent
Human PCSK9, Fc Tag (PC9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8NBP7-1). 1 * 1 mg
Catalog # ACRBPC9-H5256-1MG
Supplier:  ACROBIOSYSTEMS
Human PCSK9, Fc Tag (PC9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession Q8NBP7-1). 1 * 1 mg
Catalog # ACRBPC9-H5256-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together