Order Entry
Ireland
ContactUsLinkComponent
FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF25x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272.1). It is the FITC labeled form of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H526x) 1 * 200 µG
Catalog # ACRBMSN-HF25X200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-HF25x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272.1). It is the FITC labeled form of Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H526x) 1 * 200 µG
Catalog # ACRBMSN-HF25X200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together