Order Entry
Ireland
ContactUsLinkComponent
Human VSIG3, Fc Tag (VS3-H5258) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 241 (Accession Q5DX21-1). 1 * 1 mg
Catalog # ACRBVS3-H5258-1MG
Supplier:  ACROBIOSYSTEMS
Human VSIG3, Fc Tag (VS3-H5258) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 241 (Accession Q5DX21-1). 1 * 1 mg
Catalog # ACRBVS3-H5258-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together