Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human IL-18 R1 CD218a Protein, Fc Tag,Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK29 3). It contains AA Ala 19 - Arg 329 (Accession AAH69575).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 200 µG
Catalog # ACRBIL1-H5259200UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human IL-18 R1 CD218a Protein, Fc Tag,Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK29 3). It contains AA Ala 19 - Arg 329 (Accession AAH69575).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 200 µG
Catalog # ACRBIL1-H5259200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together