Order Entry
Ireland
ContactUsLinkComponent
Human EphA4, His Tag (EP4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Thr 547 (Accession P54764-1). 1 * 100 µG
Catalog # ACRBEP4-H52H8100UG
Supplier:  ACRO Biosystems
Human EphA4, His Tag (EP4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Thr 547 (Accession P54764-1). 1 * 100 µG
Catalog # ACRBEP4-H52H8100UG
Supplier:  ACRO Biosystems
Frequently Bought Together