Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human Apolipoprotein E APOE Protein, His Tag,Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession P02649-1).This protein carries a polyhistidine tag at t 1 * 100 µG
Catalog # ACRBAPE-H5246100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Apolipoprotein E APOE Protein, His Tag,Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession P02649-1).This protein carries a polyhistidine tag at t 1 * 100 µG
Catalog # ACRBAPE-H5246100UG
Supplier:  ACRO Biosystems
Frequently Bought Together