Order Entry
Ireland
ContactUsLinkComponent
Human FGF basic, Tag Free (BFF-H4117) is expressed from E.coli cells. It contains AA Pro 143 - Ser 288 (Accession P09038-4). 1 * 50 µG
Catalog # ACRBBFF-H4117-50UG
Supplier:  ACROBIOSYSTEMS
Human FGF basic, Tag Free (BFF-H4117) is expressed from E.coli cells. It contains AA Pro 143 - Ser 288 (Accession P09038-4). 1 * 50 µG
Catalog # ACRBBFF-H4117-50UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together