Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human TSLP (R127A, R130A), His,Avitag (TSP-H82E0) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - G ln 159 (Accession Q969D9-1 (R127A, R130A)). 1 * 200 µG
Catalog # ACRBTSP-H82E0200UG
Supplier:  ACRO Biosystems
Biotinylated Human TSLP (R127A, R130A), His,Avitag (TSP-H82E0) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - G ln 159 (Accession Q969D9-1 (R127A, R130A)). 1 * 200 µG
Catalog # ACRBTSP-H82E0200UG
Supplier:  ACRO Biosystems
Frequently Bought Together