Order Entry
Ireland
ContactUsLinkComponent
Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 1 mg
Catalog # ACRBHST-H5245-1MG
Supplier:  ACROBIOSYSTEMS
Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 1 mg
Catalog # ACRBHST-H5245-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together