Order Entry
Ireland
ContactUsLinkComponent
385013253 HWS- stirrer propeller type, with KPG 10, for DN 120 4 arms, ins stainless steel / PFA coated plane Ø 80 mm length 500 mm, connection knurled Customs No. 73269092 1 * 1 items
Catalog # HWSA385013253
Supplier:  HWS LABORTECHNIK
385013253 HWS- stirrer propeller type, with KPG 10, for DN 120 4 arms, ins stainless steel / PFA coated plane Ø 80 mm length 500 mm, connection knurled Customs No. 73269092 1 * 1 items
Catalog # HWSA385013253
Supplier:  HWS LABORTECHNIK
Frequently Bought Together