Standard solution 1000 mg/l Permanganate Index 1 * 26 mL
Catalog # HACHLCA394
Supplier: Hach
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
Electrode cleaning solutions
Supplier: Hanna
pH buffer, single-use, SINGLET™
Supplier: Hach
pH buffer solutions, colour coded
Supplier: Hach
Supelco® TraceCERT® Organic Reference Standard, Resveratrol
Supplier: Merck
Buffer solutions, colour coded, Fluka™
Supplier: Honeywell Chemicals
STANDARD S-21+K-LI-SB 100PPM 1 * 200 g
Supplier: LGC Standards PROMOCHEM
VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material
Supplier: VWR Chemicals
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution
Supplier: Thermo Fisher Scientific
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
Multi-element calibration standard 5
Supplier: PerkinElmer
Test kits, photometric, nickel, LCK cuvette tests
Supplier: Hach
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck
STANDARD AU 2MG/L IN HNO3 5% HCL 15% 1 * 500 mL
Supplier: CUSTOM MADE CHEMICALS LAB