Order Entry
Ireland
ContactUsLinkComponent
10 reactions-The Thermo Scientific SureQuant TP53 Panel Relative Quantitation Module provides a complete quantitation solution for t he analysis of key TP53 expression and phosphorylation using mass spectrometry (MS). 1 * 10 Assays
  PIERA40102
10 reactions-The Thermo Scientific SureQuant TP53 Panel Relative Quantitation Module provides a complete quantitation solution for t he analysis of key TP53 expression and phosphorylation using mass spectrometry (MS). 1 * 10 Assays
  PIERA40102
Frequently Bought Together