Order Entry
Ireland
ContactUsLinkComponent
Electronic indoor-/outdoor thermom. Dual Thermo Max/Min , -50...+70:0,1C, with temperature alarm, max/min function, PVC cable for outdoor- measurements c.3m, dimensions 65x67x23mm, outdoor probe for decelerated temperature absorbtion, in plastic tu 1 * 1 items
  AMARE906810
 :  Amarell
Electronic indoor-/outdoor thermom. Dual Thermo Max/Min , -50...+70:0,1C, with temperature alarm, max/min function, PVC cable for outdoor- measurements c.3m, dimensions 65x67x23mm, outdoor probe for decelerated temperature absorbtion, in plastic tu 1 * 1 items
  AMARE906810
 :  Amarell
Frequently Bought Together