Troemner OIML Precision Weights Weight Set(8), 50 g-1 g, OIML F2, NVLAP 1 * 1 items
OHAU30390849
: OHAUS
Troemner OIML Precision Weights Weight Set(8), 50 g-1 g, OIML F2, NVLAP 1 * 1 items
OHAU30390849
: OHAUS
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection