Order Entry
Ireland
ContactUsLinkComponent
Magnesium standard solution (1000 ppm Mg) Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 mL
Catalog # ACSD5006200
Supplier:  CPACHEM
Magnesium standard solution (1000 ppm Mg) Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 mL
Catalog # ACSD5006200
Supplier:  CPACHEM
Frequently Bought Together