Order Entry
Ireland
ContactUsLinkComponent
BioCompaact IIRF310 CG Freezer 218L, exterior stainless, refrigerant R600a, RIGHT hinged SOLID doors, wheels/skids, E-Sensor, 2 wire shelves in stainless steel 430mm, 1 wire shelf in stainless steel 300mm. 5 year warranty on cabinet. 1 * 1 items
Catalog # GRAM883520141
Supplier:  GRAM
BioCompaact IIRF310 CG Freezer 218L, exterior stainless, refrigerant R600a, RIGHT hinged SOLID doors, wheels/skids, E-Sensor, 2 wire shelves in stainless steel 430mm, 1 wire shelf in stainless steel 300mm. 5 year warranty on cabinet. 1 * 1 items
Catalog # GRAM883520141
Supplier:  GRAM
Frequently Bought Together