Order Entry
Ireland
ContactUsLinkComponent
BioCompaact IIRR310 LG Refrigerator 218L, exterior white, refrigerant R600a, RIGHT hinged SOLID doors, wheels/skids, E-Sensor, 2 wire shelves in stainless steel 430mm, 1 wire shelf in stainless steel 300mm. 5 year warranty on cabinet 1 * 1 items
  GRAM883500340
 :  GRAM
BioCompaact IIRR310 LG Refrigerator 218L, exterior white, refrigerant R600a, RIGHT hinged SOLID doors, wheels/skids, E-Sensor, 2 wire shelves in stainless steel 430mm, 1 wire shelf in stainless steel 300mm. 5 year warranty on cabinet 1 * 1 items
  GRAM883500340
 :  GRAM
Frequently Bought Together