Order Entry
Ireland
ContactUsLinkComponent
POLYPHENOLS ASSAY KIT THE METHOD RELIES ON THE TRANSFER OF ELECTRONS IN ALKALINE MEDIUM FROM PHENOLIC COMPOUNDS TO FORM A BLUE CHRO MOPHORE CONSTITUTED BY A PHOSPHOTUNGSTIC/PHOSPHOMOLYBDENUM COMPLEX 1 * 1 KIT
Catalog # GENOBAQ056
Supplier:  G-Biosciences
POLYPHENOLS ASSAY KIT THE METHOD RELIES ON THE TRANSFER OF ELECTRONS IN ALKALINE MEDIUM FROM PHENOLIC COMPOUNDS TO FORM A BLUE CHRO MOPHORE CONSTITUTED BY A PHOSPHOTUNGSTIC/PHOSPHOMOLYBDENUM COMPLEX 1 * 1 KIT
Catalog # GENOBAQ056
Supplier:  G-Biosciences
Frequently Bought Together