Sandwich High Sensitivity ELISA kit for Quantitative Detection of mouse CLEC4F. 104wells/kit, with removable strips. 1 * 1 KIT
BSBTEK2047
Sandwich High Sensitivity ELISA kit for Quantitative Detection of mouse CLEC4F. 104wells/kit, with removable strips. 1 * 1 KIT
BSBTEK2047
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection