Sandwich High Sensitivity ELISA kit for Quantitative Detection of human CRELD2. 96wells/kit, with removable strips. 1 * 1 KIT
BSBTEK1983
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human CRELD2. 96wells/kit, with removable strips. 1 * 1 KIT
BSBTEK1983
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK