Order Entry
Ireland
ContactUsLinkComponent
DRAWER (STAWA-R) FOR UB90/UB30, STAINLESS STEEL 1.4301, RAW, LOAD CAPACITY: 50 KG, VOLUME: 26 LITRE, FOR UNDERBENCH CABINETS, WITH WIDTH 1100 MM, FOR MODEL(S): UB90, UB30, DIMENSIONS W X D X H (MM): 933X444X85 1 * 1 items
Catalog # MSGA14866
Supplier:  asecos
DRAWER (STAWA-R) FOR UB90/UB30, STAINLESS STEEL 1.4301, RAW, LOAD CAPACITY: 50 KG, VOLUME: 26 LITRE, FOR UNDERBENCH CABINETS, WITH WIDTH 1100 MM, FOR MODEL(S): UB90, UB30, DIMENSIONS W X D X H (MM): 933X444X85 1 * 1 items
Catalog # MSGA14866
Supplier:  asecos
Frequently Bought Together