Order Entry
Ireland
ContactUsLinkComponent
DRAWER (STAWA-R) FOR UB90/UB30, STAINLESS STEEL 1.4301, RAW, LOAD CAPACITY: 50 KG, VOLUME: 20 LITRE, FOR UNDERBENCH CABINETS, WITH WIDTH 890/1400 MM, FOR MODEL(S): UB90, UB30, DIMENSIONS W X D X H (MM): 725X444X85 1 * 1 items
Catalog # MSGA14865
Supplier:  asecos
DRAWER (STAWA-R) FOR UB90/UB30, STAINLESS STEEL 1.4301, RAW, LOAD CAPACITY: 50 KG, VOLUME: 20 LITRE, FOR UNDERBENCH CABINETS, WITH WIDTH 890/1400 MM, FOR MODEL(S): UB90, UB30, DIMENSIONS W X D X H (MM): 725X444X85 1 * 1 items
Catalog # MSGA14865
Supplier:  asecos
Frequently Bought Together