Order Entry
Ireland
ContactUsLinkComponent
Stirrer Bearings EX, Ground joint cone made of static dissipative PTFE-EX with sealing rings on the outside to prevent sticking of t he connection and to reduce the danger of breaking glass. A special gasket made of PTFE-EX and an FKM o-ring which is 1 * 1 items
Catalog # BOHLC423-10
Supplier:  Bohlender
undefined
Stirrer Bearings EX, Ground joint cone made of static dissipative PTFE-EX with sealing rings on the outside to prevent sticking of t he connection and to reduce the danger of breaking glass. A special gasket made of PTFE-EX and an FKM o-ring which is 1 * 1 items
Catalog # BOHLC423-10
Supplier:  Bohlender
Frequently Bought Together